Anti-OSBPL5/ORP5 Rabbit Polyclonal Antibody
Catalog # ABCA_AB127171-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:KIAA1534
- Antigen Symbol:OSBPL5/ORP5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:OBPH1|ORP5|OSBPL5|Oxysterol-binding protein-related protein 5|ORP-5|OSBP-related protein 5|Oxysterol-binding protein homolog 1
- Molecular Weight:98616 Da
- Storage Temperature:Shipped at +4 °C. Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human OSBPL5 aa 200-500. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to OSBPL5/ORP5.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: OSBPL5/ORP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human