Anti-SUZ12 Rabbit Recombinant Multiclonal Antibody [clone: RP23040202]
Catalog # ABCA_AB313424-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CHET9
- Antigen Symbol:SUZ12
- Clonality:Recombinant Multiclonal
- Clone:RP23040202
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- ChIP:Yes
- Form:Liquid
- Antigen Synonyms:JJAZ1|KIAA0160|Polycomb protein SUZ12|Chromatin precipitated E2F target 9 protein|Joined to JAZF1 protein|Suppressor of zeste 12 protein homolog|ChET 9 protein
- Molecular Weight:83055 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human SUZ12
- Tested Applications:ICC/IF
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit recombinant multiclonal [RP23040202] to SUZ12.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SUZ12
Clonality: Recombinant Multiclonal
Clone: RP23040202
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;African green monkey