Anti-EIF2B2 Rabbit Polyclonal Antibody
ABCA_AB96574-50UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:EIF2BB
- Antigen Symbol:EIF2B2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:Translation initiation factor eIF2B subunit beta|S20I15|S20III15|eIF2B GDP-GTP exchange factor subunit beta
- Storage Buffer:Glycerol, Tris, Glycine
- Molecular Weight:38990 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.43 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human EIF2B2 aa 50 - 300. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to EIF2B2.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: EIF2B2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human