Anti-PDGFR alpha+PDGFR beta Rabbit Monoclonal Antibody [clone: Y92] (PE (Phycoerythrin))
Catalog # ABCA_AB307638-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD140b
- Antigen Symbol:PDGFR alpha+PDGFR beta
- Clonality:Monoclonal
- Clone:Y92
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:PDGFR|PDGFR1|PDGFRB|Platelet-derived growth factor receptor beta|PDGF-R-beta|PDGFR-beta|Beta platelet-derived growth factor receptor|Beta-type platelet-derived growth factor receptor|CD140 antigen-like family member B|Platelet-derived growth factor receptor 1|PDGFR-1
- Molecular Weight:123968 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at 4 °C. Avoid freeze/thaw cycle. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:Flow Cyt (Intra)
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
PE Rabbit monoclonal [Y92] to PDGFR alpha + PDGFR beta - C-terminal.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PDGFR alpha+PDGFR beta
Clonality: Monoclonal
Clone: Y92
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse