Anti-Sarcomeric Alpha Actinin Rabbit Recombinant Multiclonal Antibody
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1072] to Sarcomeric Alpha Actinin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Sarcomeric Alpha Actinin
Clonality: Recombinant Multiclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB314910-100U
- ABCA_AB314910-20UL
- ABCA_AB314910-10UL
- ABCA_AB314910-1ML
- Antigen Symbol:
- Sarcomeric Alpha Actinin
- Sarcomeric Alpha Actinin
- Sarcomeric Alpha Actinin
- Sarcomeric Alpha Actinin
- Antigen Name:
- Alpha-actinin-2
- Alpha-actinin-2
- Alpha-actinin-2
- Alpha-actinin-2
- Antigen Synonyms:
- Alpha-actinin skeletal muscle isoform 2|F-actin cross-linking protein|ACTN2
- Alpha-actinin skeletal muscle isoform 2|F-actin cross-linking protein|ACTN2
- Alpha-actinin skeletal muscle isoform 2|F-actin cross-linking protein|ACTN2
- Alpha-actinin skeletal muscle isoform 2|F-actin cross-linking protein|ACTN2
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Recombinant Multiclonal
- Recombinant Multiclonal
- Recombinant Multiclonal
- Recombinant Multiclonal
- Host:
- Rabbit
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 103854 Da
- 103854 Da
- 103854 Da
- 103854 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
- IHC-P
- IHC-P