Anti-CD3 zeta Armenian hamster Monoclonal Antibody [clone: H146-968] (FITC (Fluorescein Isothiocyanate))
ABCA_AB91493-50UG
: Abcam
New Product
- Size:50 µg
- Packaging:Vial
FITC Armenian hamster monoclonal [H146-968] to CD3 zeta.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD3 zeta
Clonality: Monoclonal
Clone: H146-968
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse