Anti-Histone H3 (phospho T3 + acetyl K4) Rabbit Polyclonal Antibody
Catalog # ABCA_AB272140-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:H3FA
- Antigen Symbol:Histone H3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|HIST1H3C|H3FC|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l
- Modification:phospho T3 + acetyl K4
- Molecular Weight:15404 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:0.57 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human H3C15. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Dot, ICC/IF
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Histone H3 (phospho T3, acetyl K4).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;C. elegans