Anti-Histone H3 (acetyl K9 + K14 + K18 + K23 + K27) Rabbit Polyclonal Antibody
Catalog # ABCA_AB47915-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:H3FA
- Antigen Symbol:Histone H3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- ChIP:Yes
- Form:Liquid
- Antigen Synonyms:HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|HIST1H3C|H3FC|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l
- Modification:acetyl K9 + K14 + K18 + K23 + K27
- Storage Buffer:Glycerol, Whole serum
- Molecular Weight:15404 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human H3C1 acetyl K9 + K14 + K18 + K23 + K27. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Dot
- Purification:Whole antiserum
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Histone H3 (acetyl K9 + K14 + K18 + K23 + K27).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human