Anti-Proteasome Activator Subunit 4/PSME4 Rabbit Polyclonal Antibody
Catalog # ABCA_AB157158-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:BLM10
- Antigen Symbol:Proteasome Activator Subunit 4/PSME4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:KIAA0077|PSME4|Proteasome activator complex subunit 4|Proteasome activator PA200|Protein BLM10 homolog|hBlm10
- Molecular Weight:211334 Da
- Storage Temperature:Store at 4 °C
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PSME4 aa 1-50. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Proteasome Activator Subunit 4/PSME4.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Proteasome Activator Subunit 4/PSME4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human