Anti-TIM 3 Rabbit Monoclonal Antibody [clone: EPR22285-136]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22285-136] to TIM 3 - BSA and Azide free (Capture).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TIM 3
Clonality: Monoclonal
Clone: EPR22285-136
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- ABCA_AB259759-1MG
- ABCA_AB259759-100U
- Antigen Symbol:
- TIM 3
- TIM 3
- Antigen Name:
- CD366
- CD366
- Antigen Synonyms:
- Tim3|Timd3|Havcr2|Hepatitis A virus cellular receptor 2 homolog|HAVcr-2|T-cell immunoglobulin and mucin domain-containing protein 3|T-cell immunoglobulin mucin receptor 3|T-cell membrane protein 3|TIMD-3|TIM-3
- Tim3|Timd3|Havcr2|Hepatitis A virus cellular receptor 2 homolog|HAVcr-2|T-cell immunoglobulin and mucin domain-containing protein 3|T-cell immunoglobulin mucin receptor 3|T-cell membrane protein 3|TIMD-3|TIM-3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR22285-136
- EPR22285-136
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 30934 Da
- 30934 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA