Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-160 kD Neurofilament Medium Rabbit Monoclonal Antibody [clone: EPR23951-39]
 :  Abcam
New Product
 
undefined
Anti-160 kD Neurofilament Medium Rabbit Monoclonal Antibody [clone: EPR23951-39]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCA_AB281049-1MG
  • ABCA_AB281049-100U
  • Antigen Symbol:
  • 160 kD Neurofilament Medium
  • 160 kD Neurofilament Medium
  • Antigen Name:
  • NEF3
  • NEF3
  • Antigen Synonyms:
  • NFM|NEFM|Neurofilament medium polypeptide|NF-M|160 kDa neurofilament protein|Neurofilament 3|Neurofilament triplet M protein
  • NFM|NEFM|Neurofilament medium polypeptide|NF-M|160 kDa neurofilament protein|Neurofilament 3|Neurofilament triplet M protein
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR23951-39
  • EPR23951-39
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 102472 Da
  • 102472 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C
  • Store at 4 °C
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • sELISA
  • sELISA