Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Lamin B Receptor/LBR Mouse Monoclonal Antibody [clone: BBmLBR 12.F8]
Anti-Lamin B Receptor/LBR Mouse Monoclonal Antibody [clone: BBmLBR 12.F8]
Catalog # ABCA_AB269576-100U
Supplier:  Abcam
New Product
Anti-Lamin B Receptor/LBR Mouse Monoclonal Antibody [clone: BBmLBR 12.F8]
Catalog # ABCA_AB269576-100U
Supplier:  Abcam
Supplier Number:  AB269576-100UG
New Product

About this item

Mouse monoclonal [BBmLBR 12.F8] to Lamin B Receptor/LBR - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Lamin B Receptor/LBR
Clonality: Monoclonal
Clone: BBmLBR 12.F8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse