Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SESN2/Sestrin-2 Rabbit Monoclonal Antibody [clone: EPR18907]
Anti-SESN2/Sestrin-2 Rabbit Monoclonal Antibody [clone: EPR18907]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-SESN2/Sestrin-2 Rabbit Monoclonal Antibody [clone: EPR18907]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB236025-100U
  • ABCA_AB236025-1MG
  • Antigen Symbol:
  • SESN2/Sestrin-2
  • SESN2/Sestrin-2
  • Antigen Name:
  • Hi95
  • Hi95
  • Antigen Synonyms:
  • SEST2|SESN2|Sestrin-2|Hypoxia-induced gene
  • SEST2|SESN2|Sestrin-2|Hypoxia-induced gene
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR18907
  • EPR18907
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 54494 Da
  • 54494 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C. Do not freeze.
  • Store at 4 °C. Do not freeze.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • Flow Cyt (Intra), ICC/IF
  • Flow Cyt (Intra), ICC/IF

Specifications