Anti-RBBP4 Rabbit Polyclonal Antibody
Catalog # ABCA_AB1765-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:RBAP48
- Antigen Symbol:RBBP4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Histone-binding protein RBBP4|Chromatin assembly factor 1 subunit C|Chromatin assembly factor I p48 subunit|Nucleosome-remodeling factor subunit Retinoblastoma-binding protein 4|Retinoblastoma-binding protein p48|CAF-1 subunit C|CAF-I 48 kDa subunit|CAF-I p48|RBBP-4
- Storage Buffer:Whole serum
- Molecular Weight:47656 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human RBBP4 aa 400 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Whole antiserum
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RBBP4.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RBBP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Dog