Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Calprotectin (S100A8 + S100A9) Mouse Monoclonal Antibody [clone: 270000000000]
Anti-Calprotectin (S100A8 + S100A9) Mouse Monoclonal Antibody [clone: 270000000000]
  ABCA_AB17050-50UG
 :  Abcam
New Product
undefined
Anti-Calprotectin (S100A8 + S100A9) Mouse Monoclonal Antibody [clone: 270000000000]
  ABCA_AB17050-50UG
 :  Abcam
 :  AB17050-50UG
New Product

 

  • Size:
    50 µg
  • Packaging:
    Vial

 

 

Mouse monoclonal [27E10] to Calprotectin (S100A8 + S100A9).

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Calprotectin (S100A8 + S100A9)
Clonality: Monoclonal
Clone: 270000000000
Conjugation:
Epitope: ab17050 recognises an epitope on the MRP8 + MRP14 complex which is not exposed on the individual subunits
Host: Mouse
Isotype: IgG1
Reactivity: Human