Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GOLPH4/GPP130 Rabbit Monoclonal Antibody [clone: EPR13439] (Alexa Fluor® 594)
Anti-GOLPH4/GPP130 Rabbit Monoclonal Antibody [clone: EPR13439] (Alexa Fluor® 594)
Catalog # ABCA_AB319710-100U
Supplier:  Abcam
New Product
Anti-GOLPH4/GPP130 Rabbit Monoclonal Antibody [clone: EPR13439] (Alexa Fluor® 594)
Catalog # ABCA_AB319710-100U
Supplier:  Abcam
Supplier Number:  AB319710-100UL
New Product

About this item

Alexa Fluor® 594 Rabbit monoclonal [EPR13439] to GOLPH4/GPP130.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GOLPH4/GPP130
Clonality: Monoclonal
Clone: EPR13439
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: