Anti-Prolactin Receptor/PRL-R Mouse Monoclonal Antibody [clone: B6.2]
ABCA_AB212746-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Prolactin receptor
- Antigen Symbol:Prolactin Receptor/PRL-R
- Clonality:Monoclonal
- Clone:B6.2
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PRL-R|PRLR
- Molecular Weight:69506 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Native Full Length Protein corresponding to Human PRLR
- Tested Applications:IHC-P
- Purification:Protein A/G affinity purification
- Packaging:Vial
Mouse monoclonal [B6.2] to Prolactin Receptor/PRL-R - BSA free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Prolactin Receptor/PRL-R
Clonality: Monoclonal
Clone: B6.2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human