Anti-Mitofusin 1 Mouse Monoclonal Antibody [clone: 11E91H12]
Catalog # ABCA_AB126575-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Mitofusin-1
- Antigen Symbol:Mitofusin 1
- Clonality:Monoclonal
- Clone:11E91H12
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Transmembrane GTPase MFN1|Mfn1
- Molecular Weight:83726 Da
- Storage Temperature:Store at +4 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Ammonium Sulphate Precipitation
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [11E91H12] to Mitofusin 1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Mitofusin 1
Clonality: Monoclonal
Clone: 11E91H12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse; Rat