Anti-Casein Kinase 1 alpha (phospho T321) Rabbit Polyclonal Antibody
ABCA_AB63559-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Casein kinase I isoform alpha
- Antigen Symbol:Casein Kinase 1 alpha
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:CKI-alpha|CK1|CSNK1A1
- Modification:phospho T321
- Storage Buffer:PBS, glycerol, sodium chloride
- Molecular Weight:38915 Da
- Storage Temperature:Store at −20 °C. Stable for 12 months at −20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human Casein kinase I isoform alpha phospho T321 aa 300 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to Casein Kinase 1 alpha (phospho T321).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Casein Kinase 1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human