Anti-PPP2R3B Rabbit Polyclonal Antibody
ABCA_AB72027-100UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:PPP2R3L
- Antigen Symbol:PPP2R3B
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta|PP2A subunit B isoform PR48|Protein phosphatase 2A 48 kDa regulatory subunit
- Storage Buffer:PBS, tris, citrate
- Molecular Weight:65061 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PPP2R3B. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IP
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to PPP2R3B.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PPP2R3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human