Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K20/78]
: Abcam
New Product
Mouse monoclonal [K20/78] to Kv1.1 potassium channel.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Kv1.1 potassium channel
Clonality: Monoclonal
Clone: K20/78
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse;Rat
- Catalog No:
- ABCA_AB252537-100U
- ABCA_AB252537-1MG
- Antigen Symbol:
- Kv1.1 potassium channel
- Kv1.1 potassium channel
- Antigen Name:
- Potassium voltage-gated channel subfamily A member 1
- Potassium voltage-gated channel subfamily A member 1
- Antigen Synonyms:
- MBK1|MKI|Voltage-gated potassium channel subunit Kv1.1|Kcna1
- MBK1|MKI|Voltage-gated potassium channel subunit Kv1.1|Kcna1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- K20/78
- K20/78
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P