Anti-CMPK2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB139720-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Nucleoside-diphosphate kinase
- Antigen Symbol:CMPK2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Molecular Weight:49448 Da
- Storage Temperature:Store at −20 or −80 °C. Avoid freeze/thaw cycle. Store undiluted.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human UMP-CMP kinase 2, mitochondrial. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to CMPK2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CMPK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat