Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TCP1 alpha/CCTA Rat Monoclonal Antibody [clone: 91a]
Anti-TCP1 alpha/CCTA Rat Monoclonal Antibody [clone: 91a]
Catalog # ABCA_AB240903-100U
Supplier:  Abcam
New Product
undefined
Anti-TCP1 alpha/CCTA Rat Monoclonal Antibody [clone: 91a]
Catalog # ABCA_AB240903-100U
Supplier:  Abcam
Supplier Number:  AB240903-100UG
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CCT1
  • Antigen Symbol:
    TCP1 alpha/CCTA
  • Clonality:
    Monoclonal
  • Clone:
    91a
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Antigen Synonyms:
    CCTA|TCP1|T-complex protein 1 subunit alpha|TCP-1-alpha|CCT-alpha|Chaperonin containing T-complex polypeptide 1 subunit 1
  • Molecular Weight:
    60344 Da
  • Storage Temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant Fragment Protein within Mouse Tcp1. The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    ICC/IF
  • Purification:
    Protein G affinity purification
  • Packaging:
    Vial

Specifications

About this item

Rat monoclonal [91a] to TCP1 alpha/CCTA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TCP1 alpha/CCTA
Clonality: Monoclonal
Clone: 91a
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human