Anti-CD86 Rabbit Monoclonal Antibody [clone: C86/2160R]
Catalog # ABCA_AB234401-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Clonality:Monoclonal
- Host:Rabbit
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:T-lymphocyte activation antigen CD86|Activation B7-2 antigen|B70|BU63|CTLA-4 counter-receptor B7.2|FUN-1
- Molecular Weight:37682 Da
- Storage Temperature:+4 °C
- Concentration:0.2 mg/ml
- Shipping Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Immunogen:Recombinant Full Length Protein corresponding to Human CD86.
- Tested Applications:IHC-P
- Purification:Protein A/G Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [C86/2160R] to CD86.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: C86/2160R
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human