Anti-Gankyrin Rabbit Monoclonal Antibody [clone: EPR14456(2)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR14456(2)] to Gankyrin - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Gankyrin
Clonality: Monoclonal
Clone: EPR14456(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB250947-1MG
- ABCA_AB250947-100U
- Antigen Symbol:
- Gankyrin
- Gankyrin
- Antigen Name:
- 26S proteasome non-ATPase regulatory subunit 10
- 26S proteasome non-ATPase regulatory subunit 10
- Antigen Synonyms:
- 26S proteasome regulatory subunit p28|p28(GANK)|PSMD10
- 26S proteasome regulatory subunit p28|p28(GANK)|PSMD10
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR14456(2)
- EPR14456(2)
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 24428 Da
- 24428 Da
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)