Anti-MIP-3 beta/CCL19 Rabbit Monoclonal Antibody [clone: EPR7044(2)]
: Abcam
New Product
Rabbit monoclonal [EPR7044(2)] to MIP-3 beta/CCL19.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MIP-3 beta/CCL19
Clonality: Monoclonal
Clone: EPR7044(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB192877-100U
- ABCA_AB192877-1ML
- Antigen Symbol:
- MIP-3 beta/CCL19
- MIP-3 beta/CCL19
- Antigen Name:
- ELC
- ELC
- Antigen Synonyms:
- MIP3B|SCYA19|CCL19|C-C motif chemokine 19|Beta-chemokine exodus-3|CK beta-11|Epstein-Barr virus-induced molecule 1 ligand chemokine|Macrophage inflammatory protein 3 beta|Small-inducible cytokine A19|EBI1 ligand chemokine|MIP-3-beta
- MIP3B|SCYA19|CCL19|C-C motif chemokine 19|Beta-chemokine exodus-3|CK beta-11|Epstein-Barr virus-induced molecule 1 ligand chemokine|Macrophage inflammatory protein 3 beta|Small-inducible cytokine A19|EBI1 ligand chemokine|MIP-3-beta
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR7044(2)
- EPR7044(2)
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 10993 Da
- 10993 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Concentration:
- 0.875 mg/ml
- 0.875 mg/ml
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)