Anti-JAK1+JAK2 (phospho Y1007 + Y1008) Rabbit Monoclonal Antibody [clone: E132] (HRP (Horseradish Peroxidase))
Catalog # ABCA_AB303046-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Tyrosine-protein kinase JAK2
- Antigen Symbol:JAK1+JAK2
- Clonality:Monoclonal
- Clone:E132
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Janus kinase 2|JAK-2|JAK2
- Modification:phospho Y1007 + Y1008
- Molecular Weight:130674 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Antibody labelling, Target binding affinity
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
HRP Rabbit monoclonal [E132] to JAK1 (phospho Y1034 + Y1035) + JAK2 (phospho Y1007 + Y1008).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: EPR1034Y
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: