Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Iron Total Ferrous HR Visual Test Kit 10 to 100 ppm
CHEMets® Iron Total Ferrous HR Visual Test Kit 10 to 100 ppm
Catalog # 77674-812
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Iron Total Ferrous HR Visual Test Kit 10 to 100 ppm
Catalog # 77674-812
Supplier:  CHEMetrics
Supplier Number:  K-6220D
New Product
CAS Number:  

Specifications

  • Chemical method:
    Phenanthroline (total and ferrous)
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Surface and ground water;Sea water;Drinking water

Specifications

About this item

  • Method: Phenanthroline (total and ferrous) Range: 10 to 100 ppm MDL: 10 ppm
Delivery: The K-6220D Iron (total and ferrous) HR CHEMets® test kit (10 to 100 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, 3.0 ml syringe, iron activator solution, and instructions. Note: Kit contains only enough activator solution to perform approximately 7 total iron tests.