$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Brimonidine-d4 D-Tartrate
TRC Brimonidine-d4 D-Tartrate
77978-912
:
LGC Standards
New Product
:
TRC Brimonidine-d4 D-Tartrate
77978-912
:
LGC Standards
:
TRC-B677532-1MG
New Product
:
CAS No.:
1316758-27-6
Description:
Brimonidine-d4 D-Tartrate
Size:
1 mg