CHEMets® Cyanide Visual Test Kit
Catalog # 77674-770
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Isonicotinic-Barbituric acid
- Description:Cyanide (free)
- Cat. No.:77674-770
- Application:Water analysis
- Range:0 - 0.1 ppm
0.1 - 1 ppm - No. of tests:30
Specifications
About this item
- Range: 0 to 0.1 and 0.1 to 1 ppm MDL: 0.005 ppm Method: Isonicotinic-Barbituric acid
Delivery: Kit K-3810 comes in a plastic case and contains everything needed to perform 30 tests:refill, low and high range comparators, neutralizer solution, activator solution, 25 ml sample cup, 1.0 ml syringe and instructions.