Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Hydroxy Desloratadine-d4
TRC 3-Hydroxy Desloratadine-d4
TRC 3-Hydroxy Desloratadine-d4
Catalog # 77986-387
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-Hydroxy Desloratadine-d4
Catalog # 77986-387
Supplier:
LGC Standards
Supplier Number:
TRC-H936752-1MG
New Product
CAS Number:
Specifications
CAS No.:
1246819-99-7
Description:
3-Hydroxy Desloratadine-d4
Size:
1 mg
Specifications
About this item
Product Details
Product Details