$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed Oxycodone-D6.HCl.monohydrate, 0.1 mg/ml in Methanol (as free base)
Lipomed Oxycodone-D6.HCl.monohydrate, 0.1 mg/ml in Methanol (as free base)
Lipomed Oxycodone-D6.HCl.monohydrate, 0.1 mg/ml in Methanol (as free base)
77976-108
:
LGC Standards
New Product
:
Lipomed Oxycodone-D6.HCl.monohydrate, 0.1 mg/ml in Methanol (as free base)
77976-108
:
LGC Standards
:
LPM-C-1248-HC-0.1LM
New Product
:
Matrix:
Methanol
Description:
Oxycodone-D6.HCl.monohydrate, 0.1 mg/ml in Methanol (as free base)
Size:
1 ml