CHEMets® Formaldehyde Visual Test Kit
11027-210
New Product
:
- Chemical method:Purpald
- Application:Industrial process water;Waste water;Disinfection control
- Range: 0 to 1 and 1 to10 ppm MDL: 0.1 ppm Method: Purpald
: K-4605 Kit comes in a plastic case and contains everything needed to perform 30 tests (except distilled water):refill, low and high range comparators, activator solutions, 25 ml sample cup, and instructions.