$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 3-Sulfobenzaldehyde Sodium Salt
TRC 3-Sulfobenzaldehyde Sodium Salt
77993-559
:
LGC Standards
New Product
:
TRC 3-Sulfobenzaldehyde Sodium Salt
77993-559
:
LGC Standards
:
TRC-S689165-1G
New Product
:
CAS No.:
5330-48-3
Description:
3-Sulfobenzaldehyde Sodium Salt
Size:
1 g