$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone
TRC 1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone
77982-290
:
LGC Standards
New Product
:
TRC 1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone
77982-290
:
LGC Standards
:
TRC-D425575-10MG
New Product
:
CAS No.:
1373350-57-2
Description:
1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone
Size:
10 mg