Order Entry
Puerto Rico
ContactUsLinkComponent
Cyanide Instrumental Test 0 to 0.4 ppm
Cyanide Instrumental Test 0 to 0.4 ppm
Catalog # 77674-768
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Cyanide Instrumental Test 0 to 0.4 ppm
Catalog # 77674-768
Supplier:  CHEMetrics
Supplier Number:  K-3803
New Product
CAS Number:  

Specifications

  • Chemical method:
    Isonicotinic-Barbituric acid
  • Application:
    Drinking water;Fresh surface water

Specifications

About this item

  • Range: 0 to 0.400 ppm Method: Isonicotinic-Barbituric acid
Delivery: Kit K-3803 comes in a cardboard box and contains everything needed to perform up to 29 tests (except distilled water): thirty ampoules, neutralizer solution, activator solution, 25 ml sample cup, 3.0 ml syringe, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct to readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.