$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Pyrethrin 2 (~30%, Technical Grade, Contains Pyrethin 1)
TRC Pyrethrin 2 (~30%, Technical Grade, Contains Pyrethin 1)
:
LGC Standards
New Product
TRC Pyrethrin 2 (~30%, Technical Grade, Contains Pyrethin 1)
:
LGC Standards
New Product
CAS No.:
121-29-9