Paragon Scientific Viscosity Set, Cold Crank Simulator Type: CL08, CL09, CL10, CL11, CL12, CL13, CL14, CL15, CL16, CL19, CL22, CL25(x2), CL28, CL32, CL38, CL48 and CL60
77976-034
New Product
:
- Description:Viscosity set, cold crank simulator type: CL08, CL09, CL10, CL11, CL12, CL13, CL14, CL15, CL16, CL19, CL22, CL25(x2), CL28, CL32, CL38, CL48 and CL60
- Size:18×500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component