Order Entry
Puerto Rico
ContactUsLinkComponent
Iron Comparator Round 0 to 1 ppm
Iron Comparator Round 0 to 1 ppm
Catalog # 77674-640
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Iron Comparator Round 0 to 1 ppm
Catalog # 77674-640
Supplier:  CHEMetrics
Supplier Number:  C-6001
New Product
CAS Number:  

Specifications

  • Chemical method:
    Phenanthroline (total and ferrous)
  • Cat. No.:
    77674-640
  • Increments:
    0, 0.1, 0.2, 0.3, 0.4, 0.6, 0.8, 1.0 ppm
  • Range:
    0 - 1.0 ppm

Specifications

About this item

C-6001 Low Range Iron Comparator is a component of K-6210 Iron (total and ferrous) CHEMets Visual Test Kit and K-6010 Iron (total and soluble) CHEMets Visual Kit is designed to be compared against the color formed in R-6201 Iron CHEMets Refill and R-6001 Iron CHEMets Refill. The comparator's range is applicable to many applications including surface and industrial process waters. Color standards are orange in color and cover a range of 0 to 1 ppm iron (Fe).