Paragon Scientific IP 496, Sulfur set A in light mineral oil: S @ blank, 0.01, 0.03, 0.05, 0.07, 0.1%
Catalog # 77975-822
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Light Mineral Oil
- CAS No.:7704-34-9
- Description:IP 496, Sulfur set A in light mineral oil: S @ blank, 0.01, 0.03, 0.05, 0.07, 0.1%
- Size:6×100ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065