Order Entry
Puerto Rico
ContactUsLinkComponent
 
Paragon Scientific Aniline Point, Jet Aviation Fuel (nominal value: 58.48 °C)
  77975-561
New Product
 :  
undefined
Paragon Scientific Aniline Point, Jet Aviation Fuel (nominal value: 58.48 °C)
  77975-561
 :  ALK-CRMU-APKR
New Product
 :  

 

  • Description:
    Aniline point, jet aviation fuel (nominal value: 58.48 °C)
  • Size:
    250 ml
  • Storage Temperature:
    RT

 

 

  • Shipping conditions: Room temperature