Order Entry
Puerto Rico
ContactUsLinkComponent
TRC 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
TRC 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
Catalog # 77989-299
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
Catalog # 77989-299
Supplier:  LGC Standards
Supplier Number:  TRC-M330085-25G
New Product
CAS Number:  

Specifications

  • CAS No.:
    720720-96-7
  • Description:
    5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
  • Size:
    25 g

Specifications

About this item