ContactUsLinkComponent
Iron in Brine Acidifier Solution Pack
Iron in Brine Acidifier Solution Pack
Catalog # 77674-562
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Iron in Brine Acidifier Solution Pack
Catalog # 77674-562
Supplier:  CHEMetrics
Supplier Number:  A-6001
New Product
CAS Number:  
Specifications
  • Chemical method:
    Ferric Thiocyanate (Iron in brine)
Specifications
About this item
The accessory pack supplies enough solution to perform at least 100 tests. Qty. per pack: Six 20 ml bottles.