TRC 4,5-Didehydro Brimonidine
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:151110-15-5
Specifications
Frequently Bought Together
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Cylinder Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Liner O-Rings for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Fused Silica Tubing Cutters for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tools for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Nebulizer Supplies for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Guard Cartridge Holder, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)