Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-Hydroxypyridine-2-thione Zinc
TRC 1-Hydroxypyridine-2-thione Zinc
TRC 1-Hydroxypyridine-2-thione Zinc
Catalog # 77985-849
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 1-Hydroxypyridine-2-thione Zinc
Catalog # 77985-849
Supplier:
LGC Standards
Supplier Number:
TRC-H249820-1G
New Product
CAS Number:
Specifications
CAS No.:
13463-41-7
Description:
1-Hydroxypyridine-2-thione Zinc
Size:
1 g
Specifications
About this item
Product Details
Product Details