Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed Oxycodone.HCl.monohydrate, 1 mg/ml in Methanol (as free base)
Lipomed Oxycodone.HCl.monohydrate, 1 mg/ml in Methanol (as free base)
Catalog # 77976-111
Supplier:  LGC Standards
New Product
CAS Number:  
Lipomed Oxycodone.HCl.monohydrate, 1 mg/ml in Methanol (as free base)
Catalog # 77976-111
Supplier:  LGC Standards
Supplier Number:  LPM-C-404-HC-1LM
New Product
CAS Number:  

Specifications

  • Matrix:
    Methanol
  • Description:
    Oxycodone.HCl.monohydrate, 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml

Specifications

About this item