$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Malathion beta-Monoacid
TRC Malathion beta-Monoacid
Catalog # 77988-269
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Malathion beta-Monoacid
Catalog # 77988-269
Supplier:
LGC Standards
Supplier Number:
TRC-M111020-10MG
New Product
CAS Number:
Specifications
CAS No.:
1642-51-9
Description:
Malathion beta-Monoacid
Size:
10 mg
Specifications
About this item
Product Details
Product Details