CHEMets® Oxygen, Dissolved Refill (Rhodazine D)
Catalog # 11025-138
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Rhodazine D
- Application:Boiler water;Cooling water;Sea water;Power generation;Petroleum refining;Water treatment
Specifications
About this item
This product is a refill for K-7518 dissolved oxygen CHEMets visual kit for the analysis of dissolved oxygen in water. It contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box. R-7518 dissolved oxygen CHEMets refill is compatible with C-7518 dissolved oxygen comparator.
- Method: Rhodazine D