Order Entry
Puerto Rico
ContactUsLinkComponent
TRC Galaxolide (solution 50% in diethyl phthalate)
TRC Galaxolide (solution 50% in diethyl phthalate)
Catalog # 77985-310
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC Galaxolide (solution 50% in diethyl phthalate)
Catalog # 77985-310
Supplier:  LGC Standards
Supplier Number:  TRC-G189001-5G
New Product
CAS Number:  

Specifications

  • Matrix:
    Diethyl phthalate
  • CAS No.:
    1222-05-5
  • Description:
    Galaxolide (solution 50% in diethyl phthalate)
  • Size:
    5 g

Specifications

About this item