Order Entry
Puerto Rico
ContactUsLinkComponent
Total Alkalinity Titrets® Test Kit (100 to 1000 ppm)
Total Alkalinity Titrets® Test Kit (100 to 1000 ppm)
Catalog # 75780-258
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Total Alkalinity Titrets® Test Kit (100 to 1000 ppm)
Catalog # 75780-258
Supplier:  CHEMetrics
Supplier Number:  K-9820
New Product
CAS Number:  

Specifications

  • Chemical method:
    Acid titrant with pH indicator
  • Application:
    Drinking, surface and saline water;Domestic and industrial waste water;Boiler and cooling water

Specifications

About this item

Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE

  • Range: 100 to 1000 ppm as CaCO₃ (Total) MDL: 100 ppm Method: Acid titrant with pH indicator Increments: 100, 110, 120, 130, 140, 150, 160, 180, 200, 250, 300, 350, 400, 500, 700, 1000 ppm
Delivery: The K-9820 total alkalinity Titrets® Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty Titrets®; ampoules with valve assemblies, activator solution, titrettor, 25 ml sample cup, and instructions. This test features the common calcium carbonate titration for measuring alkalinity packaged in a convenient Titrets® ampoule.